Product Name
2,4,5-Trifluoro-3-methoxybenzoyl chloride 97%
Product Number
546690
Product Brand
ALDRICH
CAS Number
112811-66-2
Molecular Weight
224.56
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
TITRATION | 96.5 - 103.5 % |
TITRATION (METHOD) | AFTER OXYGEN COMBUSTION |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |