Product Name
2′,3′-Dideoxy-3′-fluorouridine 95%
Product Number
393711
Product Brand
ALDRICH
CAS Number
41107-56-6
Molecular Weight
230.19
Test | Specification |
---|---|
Appearance (Color) | Conforms to Requirements |
White to Tan to Orange | |
Appearance (Form) | Conforms to Requirements |
Powder, Crystals, or Crystalline Powder and/or | |
Chunks | |
Infrared spectrum | Conforms to Structure |
Carbon | 44.1 - 49.8 % |
Nitrogen | 11.4 - 12.9 % |
Purity (HPLC) | ≥94.0 % |