Product Name
Tetrafluoroisophthalic acid 96%
Product Number
337668
Product Brand
ALDRICH
CAS Number
1551-39-9
Molecular Weight
238.09
Test
Specification
Appearance (Color)White to Off-White
Appearance (Form)Conforms to Requirements
 Powder or Crystals
Infrared spectrumConforms to Structure
Purity (Titration by NaOH)95.5 - 104.5 %
Purity GC VPCB≥95.5 %