Product Name
4-Bromo-2-chlorophenol 99%
Product Number
167789
Product Brand
ALDRICH
CAS Number
3964-56-5
Molecular Weight
207.45
Test | Specification |
---|---|
APPEARANCE (COLOR) | White |
APPEARANCE (FORM) | Fibers |
PURITY (GC AREA %) | ≥ 98.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |