Product Name
3,5-Dimethoxybenzaldehyde 98%
Product Number
126292
Product Brand
ALDRICH
CAS Number
7311-34-4
Molecular Weight
166.17
Test | Specification |
---|---|
APPEARANCE (COLOR) | White to Yellow |
APPEARANCE (FORM) | Powder or Crystals |
TITRATION | 97.5 - 102.5 % |
TITRATION (METHOD) | WITH HYDROXYLAMINE |
PURITY (GC AREA %) | ≥ 97.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |