Product Name
1-Diethylamino-3-butanone 98%
Product Number
126063
Product Brand
ALDRICH
CAS Number
3299-38-5
Molecular Weight
143.23
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 97.5 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |