Product Name
Ethyl 4-hydroxybenzoate ReagentPlus®, 99%
Product Number
111988
Product Brand
ALDRICH
CAS Number
120-47-8
Molecular Weight
166.17
Test
Specification
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms to Structure
Purity (GC)≥98.5 %
Solubility (Turbidity)Clear
 5% in 95% Ethanol
Solubility (Color)Colorless to Faint Yellow