Skip to Content
Merck
All Photos(1)

Key Documents

TA9H93ED72B7

Targetmol Chemicals Inc.

Isobutylshikonin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
₹20,139.63
5 MG
₹32,289.95
10 MG
₹42,612.35
25 MG
₹64,224.88

Vendor SKU: TN1137-1MG

₹20,139.63


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
1 MG
₹20,139.63
5 MG
₹32,289.95
10 MG
₹42,612.35
25 MG
₹64,224.88

About This Item

CAS Number:
Molecular Weight:
358.39

Vendor SKU: TN1137-1MG

₹20,139.63


Ships in 14 days - from Aldrich Partner.

Iupac Name

(1R)-1-(5,8-dihydroxy-1,4-dioxo-1,4-dihydro-2-naphthalenyl)-4-methyl-3-pentenyl 2-methylpropanoate

Inchi Code

1S/C20H22O6/c1-10(2)5-8-16(26-20(25)11(3)4)12-9-15(23)17-13(21)6-7-14(22)18(17)19(12)24/h5-7,9,11,16,21-22H,8H2,1-4H3/t16-/m1/s1

InChI key

BVRYLTBIGIAADD-MRXNPFEDSA-N

storage temp.

-80/-20

Purity

98.75%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Isobutylshikonin

MSDS

https://www.targetmol.com/compound/Isobutylshikonin


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service