Skip to Content
Merck
All Photos(1)

Key Documents

TA9947273092

Targetmol Chemicals Inc.

Dcimc chloride

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
₹15,692.00
500 MG
₹18,624.50
1 G
₹22,436.75

Vendor SKU: T3657-200MG

₹15,692.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
200 MG
₹15,692.00
500 MG
₹18,624.50
1 G
₹22,436.75

About This Item

CAS Number:
Molecular Weight:
290.53

Vendor SKU: T3657-200MG

₹15,692.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

3-(2,6-dichlorophenyl)-5-methyl-4-isoxazolecarbonyl chloride

Inchi Code

1S/C11H6Cl3NO2/c1-5-8(11(14)16)10(15-17-5)9-6(12)3-2-4-7(9)13/h2-4H,1H3

InChI key

IZQGELJKDARDMZ-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

98.71%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Dcimc chloride

MSDS

https://www.targetmol.com/compound/Dcimc chloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service