Skip to Content
Merck
All Photos(1)

Key Documents

SML3843

Sigma-Aldrich

BAY-985

≥98% (HPLC)

Synonym(s):

(R)-1-(4-(1-(2-(6-(6-(Dimethylamino)pyrimidin-4-yl)-1H-benzo[d]imidazol-2-ylamino)pyridin-4-yl)ethyl)piperazin-1-yl)-3,3,3-trifluoropropan-1-one, 1-(4-{(1R)-1-[2-({6-[6-(Dimethylamino)pyrimidin-4-yl]-1H-benzimidazol-2-yl}amino)-pyridin-4-yl]ethyl}piperazin-1-yl)-3,3,3-trifluoropropan-1-one, BAY 985, BAY985

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30F3N9O
CAS Number:
Molecular Weight:
553.58
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(CC(F)(F)F)N1CCN(CC1)[C@H](C)C2=CC(NC3=NC4=CC=C(C=C4N3)C5=NC=NC(N(C)C)=C5)=NC=C2

InChI

1S/C27H30F3N9O/c1-17(38-8-10-39(11-9-38)25(40)15-27(28,29)30)18-6-7-31-23(13-18)36-26-34-20-5-4-19(12-22(20)35-26)21-14-24(37(2)3)33-16-32-21/h4-7,12-14,16-17H,8-11,15H2,1-3H3,(H2,31,34,35,36)/t17-/m1/s1

InChI key

HZRJHVDNTDBTOZ-QGZVFWFLSA-N

Biochem/physiol Actions

ATP-competitive, potent and selective inhibitor against TANK-binding kinase 1 (TBK1) and IKKe.

BAY-985 is an ATP-competitive, potent and selective inhibitor against IκB kinase members TANK-binding kinase 1 (TBK1 IC50 = 2/30 nM with 10 μM/1 mM ATP) and IKK-epsilon (IKKε IC50 = 2 nM with 10 μM ATP) with 60-, 138-, 155, and ~4000-fold lower potency toward FLT3, RSK4, DRAK1, and ULK1, respectively. BAY-985 potently inhibits cellular interferon regulatory factor 3 phosphorylation (pIRF3 IC50 = 74 nM) and displays antiproliferation efficacy in human melanoma SK-MEL-2 cultures (IC50 = 900 nM). When administered orally (200 mg/kg bid. po.), BAY-985 suppresses SK-MEL-2 xengraft-derived tumor growth in mice in vivo (T/C tumor weight ratio = 0.6).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Promotion of Knee Cartilage Degradation by I?B Kinase ? in the Pathogenesis of Osteoarthritis in Human and Murine Models
Arthritis & Rheumatology (Hoboken, N.J.), 75(6), 937-949 (2023)
Julien Lefranc et al.
Journal of medicinal chemistry, 63(2), 601-612 (2019-12-21)
The serine/threonine kinase TBK1 (TANK-binding kinase 1) and its homologue IKKε are noncanonical members of the inhibitor of the nuclear factor κB (IκB) kinase family. These kinases play important roles in multiple cellular pathways and, in particular, in inflammation. Herein
Claudia Tredup et al.
ACS chemical biology, 18(4), 822-836 (2023-03-22)
Well-characterized small molecules are essential tools for studying the biology and therapeutic relevance of a target protein. However, many compounds reported in the literature and routinely studied in biomedical research lack the potency and selectivity required for mechanistic cellular studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service