Skip to Content
Merck
All Photos(1)

Key Documents

1.07881

Millipore

Standard I nutrient agar

for the cultivation of fastidious bacteria

Synonym(s):

Standard nutrient agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.5 (37 °C, 37 g/L in H2O, after autoclaving)

solubility

37 g/L

bulk density

640 kg/m3

application(s)

clinical
microbiology

suitability

Enterococcus spp.
Escherichia coli
Listeria spp.
Shigella spp.
Staphylococcus spp.
Streptococcus spp.

General description

for the cultivation of fastidious bacteria

Analysis Note

Appearance (clearness)clearAppearance (colour)yellowish-brownpH-value (25 °C)7.3 - 7.7Solidification behaviour (2 hrs., 45 °C)liquidRecovery on test medium (Staphylococcus aureus ATCC 25923 (WDCM 00034))≥ 70 %Recovery on test medium (Streptococcus pyogenes ATCC 12344)≥ 70 %Recovery on test medium (Streptococcus pneumoniae ATCC 6301)≥ 70 %Recovery on test medium (Listeria monocytogenes ATCC 19118)≥ 70 %Recovery on test medium (Enterococcus faecalis ATCC 11700)≥ 70 %Recovery on test medium (Escherichia coli ATCC 25922 (WDCM 00013))≥ 70 %Recovery on test medium (Shigella flexneri ATCC 12022 (WDCM 00126))≥ 70 %Incubation: 24 hrs.; 35 °C; aerobic.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service