LIFH324A10EF
Life Chemicals Inc.
2-bromo-5H,6H,7H-pyrrolo[1,2-a]imidazole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-bromo-6,7-dihydro-5H-pyrrolo[1,2-a]imidazole
Inchi Code
1S/C6H7BrN2/c7-5-4-9-3-1-2-6(9)8-5/h4H,1-3H2
InChI key
BMXJCILXNYCNJM-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Physical Form
solid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service