Skip to Content
Merck
All Photos(1)

Key Documents

ENA490047849

Enamine

N-(2-bromoacetyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
180
MDL number:

mp

118-121

Iupac Name

N-(2-bromoacetyl)acetamide

Inchi Code

1S/C4H6BrNO2/c1-3(7)6-4(8)2-5/h2H2,1H3,(H,6,7,8)

InChI key

OWHMEXFXNKBTMM-UHFFFAOYSA-N

storage temp.

4

Purity

91

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-249475

Melting Point

118-121

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-249475


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service