COM448620376
Combi-Blocks, Inc.
Methyl 2-(1,3-dioxoisoindol-2-yl)prop-2-enoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 2-(1,3-dioxoisoindolin-2-yl)acrylate
Inchi Code
1S/C12H9NO4/c1-7(12(16)17-2)13-10(14)8-5-3-4-6-9(8)11(13)15/h3-6H,1H2,2H3
InChI key
OSVYRWKJPFHXTJ-UHFFFAOYSA-N
storage temp.
Freezer
Purity
97%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-4079
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-4079
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service