Skip to Content
Merck
All Photos(1)

Key Documents

CIAH9885028E

ChemScene LLC (Preferred Partner)

1-Chloro-6-methoxy-isoquinoline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
193.63
MDL number:

bp

333C at 760 mmHg

mp

77C

Iupac Name

1-chloro-6-methoxyisoquinoline

Inchi Code

1S/C10H8ClNO/c1-13-8-2-3-9-7(6-8)4-5-12-10(9)11/h2-6H,1H3

InChI key

UTZJYSXOFHCSGZ-UHFFFAOYSA-N

storage temp.

4C

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/132997-77-4.html

Flash Point

155.2C

Boiling point

333C at 760 mmHg

Melting Point

77C

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-D0758.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service