Skip to Content
Merck
All Photos(1)

Key Documents

JAYH9A8ECA6E

ChemScene LLC / Jinay Pharmaceuticals Pvt Ltd. (Preferred Partner)

2-Chloro-5-iodo-4-pyridinamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
₹1,120.00
100 G
₹45,162.00

Vendor SKU: CS-W000564-1G

₹1,120.00


Ships in 5 days - from Aldrich Partner.


Select a Size

Change View
1 G
₹1,120.00
100 G
₹45,162.00

About This Item

CAS Number:
Molecular Weight:
254.46
MDL number:

Vendor SKU: CS-W000564-1G

₹1,120.00


Ships in 5 days - from Aldrich Partner.

bp

367.8C at 760 mmHg

mp

125-129C

Iupac Name

2-chloro-5-iodo-4-pyridinamine

Inchi Code

1S/C5H4ClIN2/c6-5-1-4(8)3(7)2-9-5/h1-2H,(H2,8,9)

InChI key

DEJUUKULVAIMNF-UHFFFAOYSA-N

storage temp.

4C, protect from light

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/800402-12-4.html

Flash Point

176.2C

Boiling point

367.8C at 760 mmHg

Melting Point

125-129C

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W000564.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service