Skip to Content
Merck
All Photos(1)

Key Documents

CIAH987F3BED

ChemScene LLC (Preferred Partner)

5-Isopropyl-3-methyl-1H-pyrazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
160.65
MDL number:

bp

124-126 C at 14 mmHg

mp

58-59 C

Iupac Name

3-isopropyl-5-methyl-1H-pyrazole hydrochloride

Inchi Code

1S/C7H12N2.ClH/c1-5(2)7-4-6(3)8-9-7;/h4-5H,1-3H3,(H,8,9);1H

InChI key

RWNKDXREYPDGRJ-UHFFFAOYSA-N

storage temp.

4C, sealed storage, away from moisture

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/132558-01-1.html

Boiling point

124-126 C at 14 mmHg

Melting Point

58-59 C

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0188091.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service