Skip to Content
Merck
All Photos(1)

Key Documents

CIAH987F01B6

ChemScene LLC (Preferred Partner)

6-(Trifluoromethyl)indolin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
201.15
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

289.8C at 760 mmHg

mp

184-186C

Iupac Name

6-(trifluoromethyl)-1,3-dihydro-2H-indol-2-one

Inchi Code

1S/C9H6F3NO/c10-9(11,12)6-2-1-5-3-8(14)13-7(5)4-6/h1-4,13-14H

InChI key

FMPVKTRPTXQVAK-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/1735-89-3.html

Boiling point

289.8C at 760 mmHg

Melting Point

184-186C

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W019580.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service