Skip to Content
Merck
All Photos(1)

Key Documents

ATEH988847AA

AstaTech, Inc.

MAGNESIUM 3-ETHOXY-3-OXOPROPANOATE

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
₹33,360.31
1 G
₹60,852.50
5 G
₹1,60,435.31

Vendor SKU: C15698-250MG

₹33,360.31


Ships in 17 days - from Aldrich Partner.


Select a Size

Change View
250 MG
₹33,360.31
1 G
₹60,852.50
5 G
₹1,60,435.31

About This Item

CAS Number:
Molecular Weight:
286.52
MDL number:

Vendor SKU: C15698-250MG

₹33,360.31


Ships in 17 days - from Aldrich Partner.

Iupac Name

magnesium 3-ethoxy-3-oxopropanoate

Inchi Code

1S/2C5H8O4.Mg/c2*1-2-9-5(8)3-4(6)7;/h2*2-3H2,1H3,(H,6,7);/q;;+2/p-2

InChI key

DVWUPYHFVYOPNV-UHFFFAOYSA-L

Purity

97%

Country of Origin

CN

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=156639

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C15698


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service