Skip to Content
Merck
All Photos(1)

Key Documents

ATE410295026

AstaTech, Inc.

4-(AMINOMETHYL)TETRAHYDRO-2H-PYRAN-4-CARBONITRILE HCL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176.65
MDL number:

Iupac Name

4-(aminomethyl)tetrahydro-2H-pyran-4-carbonitrile hydrochloride

Inchi Code

1S/C7H12N2O.ClH/c8-5-7(6-9)1-3-10-4-2-7;/h1-5,8H2;1H

InChI key

BWJFWIAYYYPXLG-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=8145

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=65962


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service