Skip to Content
Merck
All Photos(1)

Key Documents

APO455836908

Apollo Scientific Ltd

Methyl 3-amino-5-(4-fluorophenyl)thiophene-2-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
251.28
MDL number:

bp

153-155

Iupac Name

methyl 3-amino-5-(4-fluorophenyl)-2-thiophenecarboxylate

Inchi Code

1S/C12H10FNO2S/c1-16-12(15)11-9(14)6-10(17-11)7-2-4-8(13)5-3-7/h2-6H,14H2,1H3

InChI key

YIBRKYVKVWTHEM-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/methyl-3-amino-5-4-fluorophenylthiophene-2-carboxylate

Synonym

3-Amino-5-(4-fluorophenyl)-2-(methoxycarbonyl)thiophene, Methyl 3-amino-5-(4-fluorophenyl)-2-thenoate

Boiling point

153-155

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/methyl-3-amino-5-4-fluorophenylthiophene-2-carboxylate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service