Skip to Content
Merck
All Photos(1)

Key Documents

APO455829547

Apollo Scientific Ltd

Dimethyl 2-methoxymalonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
162.14
MDL number:

bp

12

mp

107

density

1.18

Iupac Name

dimethyl 2-methoxymalonate

Inchi Code

1S/C6H10O5/c1-9-4(5(7)10-2)6(8)11-3/h4H,1-3H3

InChI key

ORXJMBXYSGGCHG-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/dimethyl-2-methoxymalonate

Flash Point

110

Boiling point

12

Melting Point

107

density

1.18

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/dimethyl-2-methoxymalonate


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service