Skip to Content
Merck
All Photos(1)

Key Documents

AMBH990F5EF8

Ambeed, Inc.

3,3'-((2-(((Benzyloxy)carbonyl)amino)-2-((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
471.46
MDL number:

Iupac Name

3,3'-((2-(((benzyloxy)carbonyl)amino)-2-((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropionic acid

Inchi Code

1S/C21H29NO11/c23-17(24)6-9-30-13-21(14-31-10-7-18(25)26,15-32-11-8-19(27)28)22-20(29)33-12-16-4-2-1-3-5-16/h1-5H,6-15H2,(H,22,29)(H,23,24)(H,25,26)(H,27,28)

InChI key

NQLURFNELQYXEY-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Liquid or semi-solid or Solid or lump

Product Link

https://www.ambeed.com/products/200133-16-0.html

Synonym

3,3�[[2-(Cbz-amino)-2-[(2-carboxyethoxy)methyl]propane-1,3-diyl]bis(oxy)]dipropanoic Acid

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/200133-16-0.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service