Skip to Content
Merck
All Photos(1)

Key Documents

AMBH98C8E393

Ambeed, Inc.

2-Bromo-4-(4-fluorophenyl)thiazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
258.11
MDL number:

Iupac Name

2-bromo-4-(4-fluorophenyl)-1,3-thiazole

Inchi Code

1S/C9H5BrFNS/c10-9-12-8(5-13-9)6-1-3-7(11)4-2-6/h1-5H

InChI key

GSCFEPBWOIFTNA-UHFFFAOYSA-N

storage temp.

Inert atmosphere,2-8C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/412923-44-5.html

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service