Skip to Content
Merck
All Photos(3)

Documents

BMSMW9600

Benchmark SmartWasher 96 Microplate Washer for 96 Well Plates

manifolds not included, AC/DC input 115 V

Synonym(s):

Microplate washer, Washer, 96 well plate washer, wash head, cleaning, wash bottle, waste bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103200
NACRES:
NB.15

feature

manifolds not included

manufacturer/tradename

Benchmark Scientific MW9600

Looking for similar products? Visit Product Comparison Guide

General description

Research requires flexibility – the SmartWasher 96 provides it. Full plates, partial plates, and strips with U, V, C, or flat bottoms are accepted by this fully automated washer. Multiple aspiration point settings allow for minimal residual liquid in the wells after aspiration. All parameters, including dispense volume, suction time, soak and shake cycles, and shaking speed are easily programmed using the large 4.3” LCD display and keypad. Up to 100 wash protocols can be stored on board the SmartWasher 96 and recalled when needed, or transferred between machines with a flash drive. The washer comes equipped with 2 wash bottles, 1 rinse bottle and a waste bottle. All have sensors that alert the user when the solutions are low, or in the case of the waste bottle, when it needs to be emptied. An autorinse function primes the dispensing tube and wash heads to prevent cross contamination and clogs. 8 and 12channel wash heads are available separately to accommodate plates in either orientation. A transparent, acrylic shield surrounds the wash area, allowing for visualization of the plate. Should a spill or leak occur, the beveled spill well ensures that the liquid is extracted immediately to avoid crosscontamination. The SmartWasher 96 is the perfect partner for the SmartReader 96 Microplate Absorbance Reader as well as other popular readers.

Application

Washing microplates for ELISA assays

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service