Skip to Content
Merck
All Photos(1)

Documents

717029

Sigma-Aldrich

4-Bromo-2-(trimethylsilyl)thiazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10BrNSSi
CAS Number:
Molecular Weight:
236.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

liquid

density

1.384 g/mL at 25 °C

SMILES string

C[Si](C)(C)c1nc(Br)cs1

InChI

1S/C6H10BrNSSi/c1-10(2,3)6-8-5(7)4-9-6/h4H,1-3H3

InChI key

WLQGXFSBIGJXDA-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

180.0 °F - closed cup

Flash Point(C)

82.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service