Skip to Content
Merck
All Photos(1)

Key Documents

669768

Sigma-Aldrich

(1S,2S,4R)-(+)-Limonene-1,2-diol

≥97.0% (GC)

Synonym(s):

(+)-(1S,2S,4R)-Limonene glycol, (+)-1-Hydroxyneodihydrocarveol, (1S,2S,4R)-(+)-4-Isopropenyl-1-methylcyclohexan-1,2-diol, (1S,2S,4R)-(+)-p-Menth-8-en-1,2-diol, (1S,2S,4R)-8-p-Menth-8-ene-1,2-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Beilstein:
2325340
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

solid

optical activity

[α]/D +45±5°, c = 1 in acetone

SMILES string

O[C@@H]1[C@@](C)(O)CC[C@@H](C(C)=C)C1

InChI

1S/C10H18O2/c1-7(2)8-4-5-10(3,12)9(11)6-8/h8-9,11-12H,1,4-6H2,2-3H3/t8-,9+,10+/m1/s1

InChI key

WKZWTZTZWGWEGE-UTLUCORTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(1S,2S,4R)-(+)-Limonene-1,2-diol is formed during the biocatalytic oxidation of (4R)-(+)-limonene by chloroperoxidase (CPO) extracted from Caldariomyces fumago.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective oxidation of R-(+)-limonene by chloroperoxidase from Caldariomyces fumago.
Aguila S, et al.
Green Chemistry, 10(6), 647-653 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service