Skip to Content
Merck
All Photos(1)

Documents

457175

Sigma-Aldrich

Poly(epichlorohydrin-co-ethylene oxide-co-allyl glycidyl ether)

epichlorohydrin 72-77 wt. %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH(CH2Cl)CH2O]x(CH2CH2O)y[CH2CH(CH2OCH2CH=CH2)O]Z
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

contains

0.3% antioxidant

composition

epichlorohydrin, 72-77 wt. %

viscosity

63-77(100 °C, Mooney, ML 1+4)(lit.)

transition temp

Tg −36 °C

density

1.32 g/mL at 25 °C (lit.)

SMILES string

OCCO.OCC(O)CCl.OCC(O)COCC=C

Application

Oil seals, printing rollers, belting, tubing, sheet and coated fabrics.

Features and Benefits

Cured polymers have low damping and are flexible over a wide temperature range, impermeable to gases.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service