Skip to Content
Merck
All Photos(3)

Key Documents

237914

Sigma-Aldrich

Hydroquinone bis(2-hydroxyethyl) ether

98%

Synonym(s):

2,2′-(1,4-Phenylenedioxy)diethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH2CH2OH)2
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

autoignition temp.

874 °F

mp

100-102 °C (lit.)

SMILES string

OCCOc1ccc(OCCO)cc1

InChI

1S/C10H14O4/c11-5-7-13-9-1-2-10(4-3-9)14-8-6-12/h1-4,11-12H,5-8H2

InChI key

WTPYFJNYAMXZJG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evelyn C S Santos et al.
Journal of materials chemistry. B, 8(4), 703-714 (2019-12-24)
Here we describe the assembly and pH-driven operation of two nanocarriers based on non-functionalized (MCM-41) and carboxylate-functionalized (MCM-41-COOH) containers loaded with the anticancer drug doxorubicin (DOX) and capped by quaternary ammonium pillar[5]arene (P[5]A) nanogates. MCM-41 and MCM-41-COOH containers were synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service