Skip to Content
Merck
All Photos(1)

Key Documents

AC7H039C83ED

Advanced Chemical Intermediates Ltd.

2-Amino-5-methyl-benzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
139.22
MDL number:

Iupac Name

2-amino-5-methylbenzenethiol

Inchi Code

1S/C7H9NS/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,8H2,1H3

InChI key

VUMZNLOQJGKGNE-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Room temperature

Physical Form

Pale yellow solid

Product Link

https://www.acints.com/products/ACI-04795

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.acints.com/products/ACI-04795


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service