Skip to Content
Merck
All Photos(1)

Key Documents

ADVH996BD625

AChemBlock

2-Fluoroacryloyl Chloride, Solution 50wt.% in DCM

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
108.5
MDL number:

Iupac Name

2-fluoroacryloyl chloride

Inchi Code

1S/C3H2ClFO/c1-2(5)3(4)6/h1H2

InChI key

JBKRPXAZWRLPSW-UHFFFAOYSA-N

storage temp.

Store at -20 C

Purity

95%

Country of Origin

CN

Physical Form

Colorless to yellow liquid

Product Link

https://www.achemblock.com/u138931-2-fluoroacryloyl-chloride-solution-50wt-in-dcm.html

Synonym

2-fluoroacryloyl chloride

Pictograms

Signal Word

Danger

Hazard Statements

MSDS

https://www.achemblock.com/u138931-2-fluoroacryloyl-chloride-solution-50wt-in-dcm.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service