Skip to Content
Merck
All Photos(1)

Key Documents

408190

Sigma-Aldrich

2,6-Dichloro-4-(trifluoromethyl)aniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(CF3)NH2
CAS Number:
Molecular Weight:
230.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

60-62 °C/1 mmHg (lit.)

mp

34-36 °C (lit.)

density

1.532 g/mL at 25 °C (lit.)

SMILES string

Nc1c(Cl)cc(cc1Cl)C(F)(F)F

InChI

1S/C7H4Cl2F3N/c8-4-1-3(7(10,11)12)2-5(9)6(4)13/h1-2H,13H2

InChI key

ITNMAZSPBLRJLU-UHFFFAOYSA-N

Application

2,6-Dichloro-4-(trifluoromethyl)aniline may be employed for the synthesis of 1-[2,6-di­chloro-4-(tri­fluoro­methyl)­phenyl]-5-[(4-nitro­phenyl)­methyl­ene­imino]-1H-pyrazole-3-carbo­nitrile.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-[2, 6-Dichloro-4-(trifluoromethyl) phenyl]-5-[(4-nitrophenyl) methyleneimino]-1H-pyrazole-3-carbonitrile.
Zhong P, et al.
Acta Crystallographica Section E, Structure Reports Online, 61(3), 786-787 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service