Skip to Content
Merck
All Photos(1)

Key Documents

384011

Sigma-Aldrich

N,N-Diethyl-2-phenylacetamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CON(C2H5)2
CAS Number:
Molecular Weight:
191.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.522 (lit.)

bp

169-171 °C/18 mmHg (lit.)

density

1.004 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)C(=O)Cc1ccccc1

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mustapha Debboun et al.
Journal of medical entomology, 41(3), 430-434 (2004-06-10)
Seventeen analogs of the repellent compounds N,N-diethyl-3-methylbenzamide (DEET) and N,N-diethylphenylacetamide (DEPA) were evaluated in vitro for repellency against laboratory-reared Aedes aegypti (L.) and Anopheles stephensi Liston mosquitoes by using a modified membrane blood feeding test system. The system was a
Gas chromatographic identification of urinary metabolites of insect repellent N,N-diethylphenylacetamide on inhalation exposure in rats.
S S Rao et al.
Journal of chromatography, 493(1), 210-216 (1989-08-25)
R Vijayaraghavan et al.
Toxicology, 67(1), 85-96 (1991-03-25)
N,N-Diethylphenylacetamide (DEPA) is an inexpensive, long-acting and broad spectrum insect repellent. The acute LC50 for a 4-h exposure of DEPA aerosol was found to be 1.451 mg l-1 (1.290-1.633) in male and 1.375 mg l-1 (1.307-1.447) in female rats. DEPA
M V Suryanarayana et al.
Journal of pharmaceutical sciences, 80(11), 1055-1057 (1991-11-01)
A series of amide analogues of N,N-diethyl-m-toluamide (DEET) and N,N-diethylphenylacetamide (DEPA) were synthesized to study their mosquito repellency in relation to chemical structure and physical properties. None of these parameters (vapor pressure, lipophilicity, and molecular length) could be related to
M P Prasad et al.
The Indian journal of medical research, 91, 453-457 (1990-11-01)
Three controlled release formulations of the insect repellent DEPA (N, N-Diethyl phenylacetamide), Depa-A, Depa-B and Depa-C have been developed and tested on human subjects against Aedes aegypti adult mosquitoes for repellency and the results were compared to those of a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service