Merck
Skip The Scissors
Go for a safe & clean cut with Velax® cutting clamp
New Velax® Cutting Clamp for a Scissor Free Sterility Testing Workflow
Experience safety and convenience in sterility testing by using the Velax® cutting clamp, readily mounted on every Steritest® NEO device, to cut the tubing.
The Velax® cutting clamp ensures a quick and clean cut: 
  • No cross-contamination risk from scissors
  • No scissor handling and sanitization
  • Reduced safety risks for the operator
Since 2018, when we introduced the new Steritest® NEO devices, we have launched new elements every year to significantly improve your workflow safety, reliability and convenience. We are continuously raising the industry standards, and driving innovation to provide you the most secure and reliable test ever.
How to use the Velax® cutting clamp
Want to know more about the Velax® cutting clamp or get a sample?
Simply fill in & submit the form below:
How to use the Velax® cutting clamp
Media transfer step and pressure equilibration of sterility test
After the media transfer step and pressure equilibration (clamps and pump head open) of your sterility test:
  • Close the white and red clamps (on the canisters’ side)
  • Remove the tubing from the pump head
    Slide the blue Velax® cutting clamp to an area free of droplets where tubes are still joined
    Slide the blue Velax® cutting clamp to an area free of droplets where tubes are still joined (approx. 3 cm from clamps).
    Firmly press the Velax® cutting clamp as shown on the top 2 photos:
    • Both tubing lines are cut simultaneously
    • Velax® cutting clamp remains on the separated part of the tubing
      Fold the canister tubing over and insert the ends onto the vents of the canisters.
      Fold the canister tubing over and insert the ends onto the vents of the canisters. Discard the Velax® cutting clamp with tubing remains and needles.
        Related Products
        Loading
        Related Resources
        • Brochure: NEO Devices
          Thanks to our strong and long experience with the Sterility Test, we have a complete offer with the Steritest® NEO devices, the Steritest® culture media and rinsing fluids, the Steritest® Symbio pump, and broad service offering with method developement and consultancy, validation, training and more.
        • Datasheet: Steritest™ NEO Devices
          Our Steritest® NEO devices simplify every aspect of testing, from handling to traceability, all within a closed concept system.
        • Wallchart: Sterility Testing Solutions
          We invented the original Steritest® closed filtration system in the 1970’s which changed sterility testing forever.