Merck
HomePathogen & Spoilage TestingSalmonella Contamination and Rapid Detection Methods
Salmonella Contamination and Rapid Detection Methods
SALMONELLA BACTERIA
Foodborne illness affects public health and development worldwide. Salmonella spp. is a Gram-negative bacterium that is responsible for many foodborne diseases in humans and animals. Salmonella belongs to Enterobacteriaceae family and there are more than 2600 known serovars (or variations) within the Salmonella species. The serovars of Salmonella are broadly classified into typhoidal and nontyphoidal. The nontyphoidal Salmonella serovars Typhimurium and Enteritidis are two of the most common causes of gastroenteritis, but many others can also lead to illness. As Salmonella is considered a serious threat to human health, the effective identification and rapid detection of Salmonella is crucial to prevent outbreaks of food-borne disease.
Salmonella bacteria contamination and testing
INFECTION WITH SALMONELLA
Salmonellosis, the infection caused by Salmonella, can be extremely severe, particularly for young children, elderly and the immuno-compromised. It is generally contracted through the consumption of contaminated food of animal origin (mainly eggs, meat, poultry, and milk), although other foods have also been implicated in its transmission. Symptoms of Salmonellosis include diarrhea, abdominal pain, nausea, and vomiting that can last from 1 to 7 days.
SALMONELLA CONTAMINATION IN FOOD
Salmonella bacteria are widely distributed in domestic and wild animals. They are prevalent in animals such as poultry, pigs, and cattle. Salmonella can pass through the entire food chain from animal feed to primary production, and households or food-service establishments and institutions.1
Salmonella can be quite resilient, cannot be killed by freezing, and can also survive in acidic foods. The high resistance to drying, combined with a very high heat resistance once dried, makes Salmonella a potential problem in most foods, particularly in dry and semi-dry products (e.g., milk powder, spices, etc.). This means that if contamination is present, it can be difficult to eradicate. For example, fatty foods can protect the cells from quite severe heat treatments, making pasteurization ineffective.
It is therefore exceptionally important for all food manufacturers to rigorously test for Salmonella to ensure no contaminated products are released to the public.
ISO METHOD FOR SALMONELLA DETECTION
The International Organization for Standardization (ISO) recently revised the scope of the horizontal method for the detection, enumeration, and serotyping of Salmonella in the revised EN ISO 6579-1:2017 standard which has been expanded to incorporate ISO 6785 I IDF 93 and includes milk and milk products, animal feces and environmental samples from the primary production stage.
For further information, read our article ISO 6579-1 for Salmonella Detection in the Food Chain or watch our 10-minute presentation on compliant Salmonella spp. testing.
Media and ISO Compliance
When following ISO 6579-1:2017, regulatory-compliant media should be used as the media formulation and preparation should be as described in the method. For confidence during audits or lab inspections, use media that has been released from an ISO/IEC 17025 accredited laboratory, such as our Granucult® dehydrated media, or ready-to-use ReadyTube® media, which also verifies compliance with DIN EN ISO 11133:2014.
Sample Preparation and Enrichment
Sample preparation and pre-enrichment are carried out with buffered peptone water, followed by selective enrichment using Muller-Kauffmann Tetrathionate-Novobiocin (MKTTn) Broth, Rappaport Vasiliadis Soy broth (RVS) broth, or Modified Semi-solid Rappaport Vasiliadis (MSRV) agar.
Discover the new ReadyStream® system that instantly dispenses pre-heated, sterile, and ISO 11133-compliant culture media with the push of a button. The system simplifies media preparation in food testing labs and allows you to prepare up to 100 L of ready-to-use culture media in less than 20 minutes.
Check out our gravimetric dilutors and sample homogenizers to aid with sample preparation.
Selective Media for Salmonella Detection
EN ISO 6579-1:2017 offers greater flexibility in the plating-out step of Salmonella detection so the procedures for the inoculation of the isolation medium have been made less prescriptive. More detailed directions for the choice of the second isolation medium, which must be used in addition to XLD (Xylose Lysine Deoxycholate) agar, are now given. This medium must be based on different diagnostic characteristics to balance the disadvantages of XLD e.g., the lack of detection of H2S-negative Salmonella. As the second diagnostic agar, we recommend using Rambach® agar, which is based on acid formation with propylene glycol, and in combination with an included pH indicator, the colonies will have a characteristic red color. This enables species of Salmonella to be differentiated unambiguously and allows identification of H2S-negative Salmonella strains, e.g., S. Agona, which may otherwise be missed using XLD agar and a second agar based on the same diagnostic principle, such as XLT4 or Hektoen Enteric Agar.
RAPID METHODS FOR SALMONELLA DETECTION
Immunological Methods
Lateral Flow Tests are immunoassays for detecting food pathogens. Acting as mini laboratories, they include a built-in control reaction and definitive results are delivered in as little as 20 minutes, following the standard ISO 6579-1 enrichment method. Our Singlepath® Salmonella test is AOAC-RI approved.
One of the simplest rapid methods available for the detection of Salmonella uses immunodiffusion technology to provide accurate and rapid results following a simple 1 or 2-step enrichment protocol. This 1–2 Test is also an AOAC official method of analysis.
For a high-throughput method, Enzyme Immunoassay (EIA) technology using complex antibody formulations can overcome the challenges of differential bacterial detection. Extensive AOAC validation along with built-in positive and negative controls is combined to provide users with the necessary confidence in the results. Assays are also validated by AFNOR according to ISO 16140.
Molecular Methods
Try our rapid and accurate detection method featuring the latest innovations in genetic detection technology and food microbiology. The Assurance® GDS system uses multiple layers of specificity, including immunomagnetic separation (IMS), highly specific primers, and a patented probe to ensure highly accurate results. GDS assays have been validated by AOAC and other international standards (AFNOR/MicroVal) according to ISO 16140.
Reference Materials for QC and Validation
To ensure the quality of your test results, it is advisable to use certified reference microorganisms for verification. Our Vitroids™  and LENTICULE® discs contain viable microorganisms in a certified quantity (generally accredited according to ISO/IEC 17025) and are produced under reproducible conditions compliant with ISO Guide 34:2009 using authenticated strains from NCTC®, NCPF®, and CECT®. These discs consist of pure cultures of bacteria in a solid water-soluble matrix, that are stable for at least one year and available in a viable state with a shelf life of 1-3 years. Each batch is provided with a comprehensive certificate of analysis (CoA).
Materials
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
WHO Salmonella (non-typhoidal) Fact Sheet.. [Internet].[cited 22 Jan 2018]. Available from: http://www.who.int/mediacentre/factsheets/fs139/en/