Merck
LC/MS Analysis of Sorbitol and Mannitol on SeQuant® ZIC-HILIC
LC/MS Analysis of Sorbitol and Mannitol on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-cHILIC, PEEK, 250 x 4.6 mm, 3 μm, 100Å (1.50662.0001)
mobile phase
[A] acetonitrile; [B] 0.1% ortho-phosphoric acid in Milli-Q® water; (90:10, A:B)
flow rate
1.5 mL/min
pressure
1682 psi (116 bar)
column temp.
45 °C (oven), 40 °C (detector cell)
detector
Refractive Index (RI) (cell: 10 μL)
injection
20 μL
sample
5.0 mg/mL (5000 ppm) each of sorbitol and mannitol in diluent. (Diluent: Acetonitrile: 0.1% ortho-phosphoric acid in Milli-Q® water 75:25 (v/v))
Description
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany