Merck
LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide
LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 10 mM ammonium acetate (pH 5.0 with acetic acid): [B] acetonitrile; (20:80, A:B)
flow rate
1 mL/min, split to the MS
column temp.
35 °C
detector
MS ESI (+), SIR Mode
injection
5 μL
sample
1 μg/mL in water:acetonitrile (1:1)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany