HPLC Analysis of Xylose, Fructose, Glucose and Saccharose on Chromolith® NH2 100 - 4.6mm
CONDITIONS
column
Chromolith® NH2 100 x 4.6mm (1.52028.0001)
mobile phase
[A] acetonitrile; [B] water; (75:25, A:B)
flow rate
1.0 mL/min
pressure
160 psi (11 bar)
column temp.
23 °C ambient
detector
190 nm cell: 11 μL
injection
2 μL
sample
Xylose 2.28 mg/mL, Fructose 6.45 mg/mL, Glucose 9.20 mg /mL, and Saccharose 9.42 mg/mL diluted in mobile phase.
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany