HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Mobile Phase pH
![HPLC Analysis of mAb-X22 on Antibodix® WCX-NP10, Impact of Mobile Phase pH application for HPLC](/deepweb/assets/sigmaaldrich/product/images/414/090/ed386ac9-725a-4c74-9166-c10bf2020c5f/800/ed386ac9-725a-4c74-9166-c10bf2020c5f.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase
(A) 10 mM sodium phosphate buffer at 7.5 pH; (B), A + 100 mM NaCl at 7.0 pH
gradient
15 to 65% B in 60 min
flow rate
0.8 mL/min
column temp.
25 °C
detector
UV, 214 nm
injection
10 μL
sample
MAb-X22, 1.5 mg/mL
Description
Legal Information
Antibodix is a registered trademark of Sepax Technologies