Merck
HPLC Analysis of Four Proteins on Zenix® SEC-300, Comparison of Column Internal Diameter
HPLC Analysis of Four Proteins on Zenix® SEC-300, Comparison of Column Internal Diameter application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 2.1 mm, 3 μm particle size, stainless steel
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 4.6 mm, 3 μm particle size, stainless steel
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
standard
Product No.
Description
Pricing
Myoglobin from equine heart
≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
IgG from human serum
reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Uracil
≥99.0%
Thyroglobulin from bovine thyroid
(For Gel Filtration Chromatography)
Bovine Serum Albumin
lyophilized powder, ≥96% (agarose gel electrophoresis)
CONDITIONS
column
Zenix SEC-300, 30 cm x 2.1 mm I.D., 3 μm particles (Z777024)
column
Zenix SEC-300, 30 cm x 4.6 mm I.D., 3 μm particles (Z777028)
column
Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777033)
mobile phase
0.15 M potassium phosphate monobasic, pH 7.0 with phosphoric acid
flow rate
15, 72, 207 μL/min (2.1 mm ID, 4.6 mm ID, 7.8 mm ID, respectively)
pressure
220, 240, 350 psi (2.1 mm ID, 4.6 mm ID, 7.8 mm ID, respectively)
column temp.
25 °C
detector
UV, 215 nm
injection
0.3, 1.4, 4.0 μL (2.1 mm ID, 4.6 mm ID, 7.8 mm ID, respectively)
sample
(Waters SEC200 Protein Std)
Description
Analysis Note
Smaller column internal diameters (ID) are attractive for reasons of low solvent consumption. However, when adequate sample is available, larger ID analytical columns can be advantageous because they provide better column performance.
Legal Information
Zenix is a registered trademark of Sepax Technologies