Merck
HPLC Analysis of Chinese Ginseng Extract on Ascentis® Express C18
HPLC Analysis of Chinese Ginseng Extract on Ascentis® Express C18 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] water; [B] acetonitrile
gradient
held at 25% B for 1.5 min; 25 to 85% B in 12 min; held at 85% for 1 min
flow rate
1 mL/min
pressure
3481 psi (240 bar)
column temp.
60 °C
detector
UV, 205 nm
injection
25 μL
sample
Extracted from dried, ground plant material; 10 mg/mL in water:acetonitrile (85:15); sonicate 10 min at ambient
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of Chinese ginseng root extract.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany