HPLC Analysis of Antipyretics/Analgesics/Antifungals on Discovery® RP-AmideC16
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery RP-AmideC16, 15 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
0.1% TFA in (25:75) acetonitrile:water
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany