Merck
HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® TAG
HPLC Analysis of Alanine Enantiomers on Astec® CHIROBIOTIC® TAG application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® TAG Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
DL-Alanine
≥99% (HPLC)
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
[A] water; [B] acetonitrile; (50:50, A:B)
flow rate
0.8 mL/min
column temp.
25 °C
detector
UV, 210 nm
injection
2 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC