Merck
DART®-MS Analysis of Drugs of Abuse in Human Urine Using C18 SPME LC (BioSPME) Tips
DART®-MS Analysis of Drugs of Abuse in Human Urine Using C18 SPME LC (BioSPME) Tips application for HPLC, application for SPME
Materials
SPME fiber
Product No.
Description
Pricing
SPME LC Tips, C18
tray of 96 ea
standard
Product No.
Description
Pricing
(±)-Methadone solution
1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Cocaine-D3 solution
100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Cocaine solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Cocaethylene-D3 solution
100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Cocaethylene solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
EDDP-D3 perchlorate solution
100 μg/mL in methanol (as pyrrolinium), ampule of 1 mL, certified reference material, Cerilliant®
EDDP perchlorate solution
1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®
(±)-Methadone-D3 solution
100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
sample/matrix
human urine
SPME fiber
SPME LC Tips C18 (57234-U)
extraction
60 min with 1 mL sample
desorption process
Direct MS
detector
MS, Full scan 100-500 amu
Description
Analysis Note
Economical and user-friendly sample prep method for DART-MS detection and identification of drugs of abuse in urine.
Legal Information
DART is a registered trademark of JEOL USA, Inc.