Merck
GC Analysis of FAMEs in Palm Oil on SLB-IL111
GC Analysis of FAMEs in Palm Oil on SLB-IL111 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL111 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
oven
180 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 25 cm/sec
injection
1 μL, 50:1 split
liner
4 mm I.D., split type, cup design
sample
Characterized reference oils, methylated using BF3-methanol
prior to analysis
column
SLB-IL111, 30 m x 0.25 mm I.D., 0.20 μm (28927-U)