GC Analysis of Enantiomers on γ-DEX™ 120
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
γ-DEX 120, 30 m × 0.25 mm I.D., 0.25 μm (24307)
oven
90 °C, 1 °C/min to 135 °C, 5 °C/min to 240 °C
inj. temp.
300 °C
detector
FID, 300 °C
carrier gas
helium, 20 cm/sec
injection
1 μL 100:1 split
sample
500 μg/mL in methylene chloride
Description
Legal Information
DEX is a trademark of Sigma-Aldrich Co. LLC