GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8
Materials
SPE tube or plate
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
sample/matrix
waste water spiked with pesticides
SPE tube/cartridge
Supelclean ENVI-8, glass hardware, 500 mg/6 mL (57107 (500 mg/6 mL); 57108 (1 g/6 mL))
condition
3 mL methanol; h 2 mL 5% methanol in water
sample addition
load sample at 5 mL/min
drying
2-3 min with nitrogen stream
elution
2 x 4 mL hexane:acetone (90:10), drop wise. allow to soak into bed
column
5% Phenyl Polysiloxane, 15 m × 0.53 mm I.D., 0.50 μm (Supelco equivalent, Equity-5, 28252-U)
oven
150 °C (0.5 min) to 275 °C (5 min)
inj. temp.
200 °C
detector
ECD, 300 °C
carrier gas
helium
injection
1 μL split/splitless (45 sec delay)
Description
Application
Sample Pre-treatment:
Add 5 mL methanol to 100 mL aqueous hazardous waste. Adjust to pH 5-7 if necessary.
Legal Information
ENVI is a trademark of Sigma-Aldrich Co. LLC
Supelclean is a trademark of Sigma-Aldrich Co. LLC