GC Analysis of FAMEs in Canola Oil on SLB-IL111
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL111, 30 m x 0.25 mm I.D., 0.20 μm (28927-U)
oven
(180 °C)
inj. temp.
(250 °C)
detector
(FID, 260 °C)
injection
(1 μL, 50:1 split)
liner
(4 mm I.D., split type, cup design)
sample
(Canola oil characterized reference oil (46961), methylated using BF3-methanol prior to analysis)
carrier gas
(helium, 25 cm/sec)