Merck
  • Identification and quantification of seven volatile n-nitrosamines in cosmetics using gas chromatography/chemical ionization-mass spectrometry coupled with head space-solid phase microextraction.
Identification and quantification of seven volatile n-nitrosamines in cosmetics using gas chromatography/chemical ionization-mass spectrometry coupled with head space-solid phase microextraction.
Talanta (2015-12-15)
Na Rae Choi, Yong Pyo Kim, Won Hyun Ji, Geum-Sook Hwang, Yun Gyong Ahn
ABSTRACT
An analytical method was developed for the identification and quantification of seven volatile n-nitrosamines (n-nitrosodimethylamine [NDMA], n-nitrosoethylmethylamine [NMEA], n-nitrosodiethylamine [NDEA], n-nitrosodipropylamine [NDPA], n-nitrosodibutylamine [NDBA], n-nitrosopiperidine [NPIP], and n-nitrosopyrrolidine [NPYR]) in water insoluble cream type cosmetics. It was found that the head space-solid phase microextraction (HS-SPME) was suitable for extraction, clean up, and pre-concentration of n-nitrosamines in the cream type samples so its optimal conditions were investigated. Identification and quantification of n-nitrosamines using single quadrupole gas chromatography/mass spectrometry (GC/MS) in chemical ionization (CI) mode were carried out with accurate mass measurements. Their accurate masses of protonated molecular ions were obtained within 10 mDa of the theoretical masses when sufficiently high signal was acquired from the unique calibration method using mass and isotope accuracy. For the method validation of quantification, spiking experiments were carried out to determine the linearity, recovery, and method detection limit (MDL) using three deuterated internal standards. The average recovery was 79% within 20% relative standard deviation (RSD) at the concentration of 50 ng/g. MDLs ranged from 0.46 ng/g to 36.54 ng/g, which was satisfactory for the directive limit of 50 ng/g proposed by the European Commission (EC). As a result, it was concluded that the method could be provided for the accurate mass screening, confirmation, and quantification of n-nitrosamines when applied to cosmetic inspection.
Product Description
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Sampling Stand, for use with 4 mL vials
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Sampling Stand, for use with 15 mL vials
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€146.00
25 g
Please contact Customer Service for Availability
€313.00