Merck
  • Irf8 regulates the progression of myeloproliferative neoplasm-like syndrome via Mertk signaling in zebrafish.
Irf8 regulates the progression of myeloproliferative neoplasm-like syndrome via Mertk signaling in zebrafish.
Leukemia (2017-06-20)
F Zhao, Y Shi, Y Huang, Y Zhan, L Zhou, Y Li, Y Wan, H Li, H Huang, H Ruan, L Luo, L Li
ABSTRACT
Interferon regulatory factor (IRF)-8 is a critical transcription factor involved in the pathogenesis of myeloid neoplasia. However, the underlying mechanisms in vivo are not well known. Investigation of irf8-mutant zebrafish in this study indicated that Irf8 is evolutionarily conserved as an essential neoplastic suppressor through tight control of the proliferation and longevity of myeloid cells. Surviving irf8 mutants quickly developed a myeloproliferative neoplasm (MPN)-like disease with enhanced output of the myeloid precursors, which recurred after transplantation. Multiple molecules presented notable alteration and Mertk signaling was aberrantly activated in the hematopoietic cells in irf8 mutants. Transgenic mertk overexpression in Tg(coro1a:mertk) zebrafish recapitulated the myeloid neoplasia-like syndrome in irf8 mutants. Moreover, functional interference with Mertk, via morpholino knockdown or genetic disruption, attenuated the myeloid expansion phenotype caused by Irf8 deficiency. Therefore, Mertk signaling is a critical downstream player in the Irf8-mediated regulation of the progression of myeloid neoplasia. Our study extends the understanding of the mechanisms underlying leukemogenesis.
Sigma-Aldrich
Sudan Black B Staining System
Pricing and availability is not currently available.